Merck
The effect of vinyl acetate in acetoclastic methanogenesis.
Bioresource technology (2010-10-12)
U Durán, J Gómez, O Monroy, F Ramírez
RESUMEN
The influence of vinyl acetate (VA) in the methanogenesis was evaluated, by using an upflow anaerobic sludge blanket reactor of 1.5L. The reactor was operated at 33.5 g/L volatile suspended solids to 30±2 °C, a hydraulic residence time of 1 day, an organic loading rate of 1 kgCOD/m3/d of two different mixtures of VA and glucose. The VA was methanized to 81% when its proportion was of 10% into reactor loading rate, when VA proportion increased to 25%, the methane production rate decreased to 62% and the acetate production rate increased almost 8 times. These results indicated that VA was only hydrolyzed and glucose was not used as a co-substrate. The effect of glucose on VA methanogenic degradation was evaluated through batch reactors of 60 mL, concluding that the glucose supported the methanogenesis without favoring the VA elimination. On the other hand, the results of the sludge from the reactor in the presence of VA demonstrated that VA caused an irreversibly inhibition of acetoclastic methanogenesis when the anaerobic sludge was exposed to this compound.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Vinyl acetate, contains 3-20 ppm hydroquinone as inhibitor, ≥99%
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Supelco
Vinyl acetate, analytical standard
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Supelco
Vinyl acetate solution, certified reference material, 5000 μg/mL in acetonitrile
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Vinyl acetate, European Pharmacopoeia (EP) Reference Standard
En este momento no podemos mostrarle ni los precios ni la disponibilidad