Merck
  • Whole influenza virus vaccine is more immunogenic than split influenza virus vaccine and induces primarily an IgG2a response in BALB/c mice.
Whole influenza virus vaccine is more immunogenic than split influenza virus vaccine and induces primarily an IgG2a response in BALB/c mice.
Scandinavian journal of immunology (2005-08-12)
A-O Hovden, R J Cox, L R Haaheim
RESUMEN
The aim of this study was to compare the kinetics and the magnitude of the humoral immune response to two different influenza vaccine formulations, whole and split virus vaccines. BALB/c mice were immunized intramuscularly with one or two doses (3 weeks apart) of 7.5, 15 or 30 microg of haemagglutinin of monovalent A/Panama/2007/99 (H3N2) split or whole virus vaccine. The two vaccine formulations induced similar kinetics of the antibody-secreting cells response; however, differences in the magnitude were observed in the spleen and bone marrow. Vaccination with whole virus vaccine generally elicited a quicker and higher neutralizing antibody response, particularly after the first dose of vaccine. The two vaccine formulations gave different immunoglobulin G (IgG) subclass profiles. Split virus vaccine stimulated both IgG1 and IgG2a antibodies suggestive of mixed T-helper 1 (Th1) and Th2 response, whereas whole virus vaccine induced mainly an IgG2a antibody response, which is indicative of a dominant Th1 response. The increased immunogenicity of whole virus vaccine in a naïve population could reduce the vaccine concentration needed to provide protective immunity.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
IgM, Kappa from murine myeloma, clone TEPC 183, purified immunoglobulin, buffered aqueous solution