Merck
Transient expression of the β-glucuronidase gene in Cannabis sativa varieties.
Plant signaling & behavior (2020-06-20)
Aleksei Sorokin, Narendra Singh Yadav, Daniel Gaudet, Igor Kovalchuk
RESUMEN
In plant biology, transient expression analysis plays a vital role to provide a fast method to study the gene of interest. In this study, we report a rapid and efficient method for transient expression in Cannabis sativa seedlings using Agrobacterium tumefaciens-mediated transformation. A. tumefaciens strain EHA105 carrying the pCAMBIA1301 construct with uidA gene was used to transform cannabis seedlings and the GUS assay (a measurement of β-glucuronidase activity) was used to detect the uidA expression. In the current study, we have also established a rapid germination protocol for cannabis seeds. The all three steps seed sterilization, germination and seedlings development were carried out in a 1% H2O2 solution. Transient transformation revealed that both cotyledons and young true leaves are amenable to transformation. Compared with tobacco (Nicotiana benthamiana), cannabis seedlings were less susceptible to transformation with A. tumefaciens. Susceptibility to Agrobacterium transformation also varied with the different cannabis varieties. The method established in this study has the potential to be an important tool for gene function studies and genetic improvement in cannabis.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Sacarosa, for molecular biology, ≥99.5% (GC)
Número de referencia del producto (SKU)
Tamaño de envase
Disponibilidad
Precio
Cantidad
Sigma-Aldrich
Potassium hexacyanoferrate(II) trihydrate, ACS reagent, 98.5-102.0%
Número de referencia del producto (SKU)
Tamaño de envase
Disponibilidad
Precio
Cantidad
Sigma-Aldrich
MES, low moisture content, ≥99% (titration)
Número de referencia del producto (SKU)
Tamaño de envase
Disponibilidad
Precio
Cantidad
Sigma-Aldrich
Potassium ferricyanide(III), powder or chunks, <10 μm, 99%
Número de referencia del producto (SKU)
Tamaño de envase
Disponibilidad
Precio
Cantidad
Sigma-Aldrich
Rifampicina, ≥95% (HPLC), powder or crystals
Número de referencia del producto (SKU)
Tamaño de envase
Disponibilidad
Precio
Cantidad
Sigma-Aldrich
3′,5′-Dimethoxy-4′-hydroxyacetophenone, 97%
Número de referencia del producto (SKU)
Tamaño de envase
Disponibilidad
Precio
Cantidad
Sigma-Aldrich
Triton X-100, laboratory grade
Número de referencia del producto (SKU)
Tamaño de envase
Disponibilidad
Precio
Cantidad
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Número de referencia del producto (SKU)
Tamaño de envase
Disponibilidad
Precio
Cantidad