Merck
A Bartonella Effector Acts as Signaling Hub for Intrinsic STAT3 Activation to Trigger Anti-inflammatory Responses.
Cell host & microbe (2020-02-27)
Isabel Sorg, Christoph Schmutz, Yun-Yueh Lu, Katja Fromm, Lena K Siewert, Alexandra Bögli, Kathrin Strack, Alexander Harms, Christoph Dehio
RESUMEN
Chronically infecting pathogens avoid clearance by the innate immune system by promoting premature transition from an initial pro-inflammatory response toward an anti-inflammatory tissue-repair response. STAT3, a central regulator of inflammation, controls this transition and thus is targeted by numerous chronic pathogens. Here, we show that BepD, an effector of the chronic bacterial pathogen Bartonella henselae targeted to infected host cells, establishes an exceptional pathway for canonical STAT3 activation, thereby impairing secretion of pro-inflammatory TNF-α and stimulating secretion of anti-inflammatory IL-10. Tyrosine phosphorylation of EPIYA-related motifs in BepD facilitates STAT3 binding and activation via c-Abl-dependent phosphorylation of Y705. The tyrosine-phosphorylated scaffold of BepD thus represents a signaling hub for intrinsic STAT3 activation that is independent from canonical STAT3 activation via transmembrane receptor-associated Janus kinases. We anticipate that our findings on a molecular shortcut to STAT3 activation will inspire new treatment options for chronic infections and inflammatory diseases.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Medio RPMI-1640, With L-glutamine and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Tris(2-carboxietil)fosfina hydrochloride, powder
Sigma-Aldrich
L-Glutamina solution, 200 mM, solution, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
Yodoacetamida, BioUltra
Sigma-Aldrich
N-Acetyl-L-cysteine, BioXtra, ≥99% (TLC)