Merck
  • Kinesin-2 from C. reinhardtii Is an Atypically Fast and Auto-inhibited Motor that Is Activated by Heterotrimerization for Intraflagellar Transport.
Kinesin-2 from C. reinhardtii Is an Atypically Fast and Auto-inhibited Motor that Is Activated by Heterotrimerization for Intraflagellar Transport.
Current biology : CB (2020-03-07)
Punam Sonar, Wiphu Youyen, Augustine Cleetus, Pattipong Wisanpitayakorn, Sayed I Mousavi, Willi L Stepp, William O Hancock, Erkan Tüzel, Zeynep Ökten
RESUMEN
Construction and function of virtually all cilia require the universally conserved process of intraflagellar transport (IFT) [1, 2]. During the atypically fast IFT in the green alga C. reinhardtii, on average, 10 kinesin-2 motors "line up" in a tight assembly on the trains [3], provoking the question of how these motors coordinate their action to ensure smooth and fast transport along the flagellum without standing in each other's way. Here, we show that the heterodimeric FLA8/10 kinesin-2 alone is responsible for the atypically fast IFT in C. reinhardtii. Notably, in single-molecule studies, FLA8/10 moved at speeds matching those of in vivo IFT [4] but additionally displayed a slow velocity distribution, indicative of auto-inhibition. Addition of the KAP subunit to generate the heterotrimeric FLA8/10/KAP relieved this inhibition, thus providing a mechanistic rationale for heterotrimerization with the KAP subunit fully activating FLA8/10 for IFT in vivo. Finally, we linked fast FLA8/10 and slow KLP11/20 kinesin-2 from C. reinhardtii and C. elegans through a DNA tether to understand the molecular underpinnings of motor coordination during IFT in vivo. For motor pairs from both species, the co-transport velocities very nearly matched the single-molecule velocities, and both complexes spent roughly 80% of the time with only one of the two motors attached to the microtubule. Thus, irrespective of phylogeny and kinetic properties, kinesin-2 motors work mostly alone without sacrificing efficiency. Our findings thus offer a simple mechanism for how efficient IFT is achieved across diverse organisms despite being carried out by motors with different properties.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Tris(2-carboxietil)fosfina hydrochloride, powder
Sigma-Aldrich
D-(+)-Glucosa, ≥99.5% (GC)
Millipore
Gel ANTI-FLAG® M2 Affinity, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
TWEEN® 20, for molecular biology, viscous liquid
Sigma-Aldrich
Ammonium bicarbonate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Adenosina 5′-trifosfato disodium salt hydrate, Grade I, ≥99%, from microbial
Sigma-Aldrich
PIPES, ≥99% (titration)
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
DL-Ditiotreitol, ≥98% (HPLC), ≥99.0% (titration)
Sigma-Aldrich
Catalase from bovine liver, aqueous solution, ≥30,000 units/mg protein
Sigma-Aldrich
Sodium acetate, anhydrous, for molecular biology, ≥99%