Merck
High-throughput screening identified mitoxantrone to induce death of hepatocellular carcinoma cells with autophagy involvement.
Biochemical and biophysical research communications (2019-10-28)
Bushan Xie, Xingxing He, Guihai Guo, Xiao Zhang, Jinping Li, Jianping Liu, Yingbo Lin
RESUMEN
The use of highly efficient high-throughput screening (HTS) platform has recently gained more attention as a plausible approach to identify de novo therapeutic application potential of conventional anti-tumor drugs for cancer treatments. In this study, we used hepatocellular carcinoma (HCC) cells as models to identify cytotoxic compounds by HTS. To identify cytotoxic compounds for potential HCC treatments, 3271 compounds from three well established small molecule libraries were screened against HCC cell lines. Thirty-two small molecules were identified from the primary screen to induce cell death. Particularly, mitoxantrone (MTX), which is an established antineoplastic drug, significantly and specifically inhibited the growth and proliferation of HCC cells in vitro. Mechanistic studies of LC3-II, p62 and phosphorylation of p70S6K in HepG2 cells revealed that MTX treatment induced mTOR-dependent autophagy activation, which was further confirmed by the autophagic flux assay using lysosomal inhibitor chloroquine (CQ). In the combined treatment of MTX and CQ, where autophagy was inhibited by CQ, the elevations of cleaved Caspase-3 and PARP were observed, indicating the enhanced apoptosis in HepG2 cells. Taken together, we hypothesize that MTX-induced autophagy plays an pro-survival role in HCC treatment. Combined treatment with autophagy inhibitor may combat the chemo-resistance of HCC to MTX treatment and therefore deserves future clinical investment.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Chloroquine diphosphate salt, powder or crystals, 98.5-101.0% (EP)
Número de referencia del producto (SKU)
Tamaño de envase
Disponibilidad
Precio
Cantidad
Sigma-Aldrich
Mitoxantrone dihydrochloride, ≥97% (HPLC)
Número de referencia del producto (SKU)
Tamaño de envase
Disponibilidad
Precio
Cantidad
Sigma-Aldrich
bisBenzimide H 33258, powder, BioReagent, suitable for cell culture, ≥98% (HPLC and TLC)
Número de referencia del producto (SKU)
Tamaño de envase
Disponibilidad
Precio
Cantidad