Merck
Kallistatin Inhibits Atherosclerotic Inflammation by Regulating Macrophage Polarization.
Human gene therapy (2018-09-13)
Bing Li, Zulong Sheng, Chang Liu, Linglin Qian, Yuehuan Wu, Yanping Wu, Genshan Ma, Yuyu Yao
RESUMEN
Kallistatin (KS) has been recognized as a plasma protein with anti-inflammatory functions. Macrophages are the primary inflammatory cells in atherosclerotic plaques. However, it is unknown whether KS plays a role in macrophage development and the pathogenesis of atherosclerosis. This study investigated the role of KS in macrophage development, a key pathological process in atherosclerosis. An atherosclerosis model was established in ApoE-/- mice via partial left carotid artery (PLCA) ligation. An adenovirus vector (Ad. HKS) containing the human KS gene was delivered via the tail vein before PLCA ligation. The mice were divided into two groups: the PLCA + Ad. HKS and PLCA + adenovirus vector (Ad. Null) groups and followed for 2 and 4 weeks. Human KS was expressed in the mice after KS gene delivery. In addition, KS significantly inhibited plaque formation and reduced inflammation in the plaques and liver 4 weeks after gene delivery. Moreover, KS gene delivery significantly increased the expression of interleukin-10 and Arginase 1, which are M2 macrophage markers, and reduced the expression of inducible nitric oxide synthase and monocyte chemotactic protein 1, which are M1 macrophage markers. Furthermore, in cultured RAW 264.7 macrophages, KS significantly stimulated M2 marker expression and differentiation and decreased M1 marker expression, as determined by flow cytometry and real-time polymerase chain reaction. These effects were blocked by Krüppel-like factor 4 small-interfering RNA oligonucleotides. These findings demonstrate that KS inhibits atherosclerotic plaque formation and regulates M1/M2 macrophage polarization via Krüppel-like factor 4 activation.