Merck
Purification and characterization of a phosphoprotein phosphatase from bovine adrenal cortex.
Biochimica et biophysica acta (1975-10-22)
B Ullman, R L Perlman
PMID241403
RESUMEN
A phosphoprotein phosphatase which is active against chemically phosphorylated protamine has been purified about 500-fold from bovine adrenal cortex. The enzyme has a pH optimum between 7.5 and 8.0, and has an apparent Km for phosphoprotamine of about 50 muM. The hydrolysis of phosphoprotamine is stimulated by salt, and by Mn2+. Hydrolysis of phosphoprotamine is inhibited by ATP, ADP, AMP, and Pi, but is not affected by AMP or cyclic GMP. The purified phosphoprotein phosphatase preparation also dephosphorylates p-nitrophenyl phosphate and phosphohistone, and catalyzes the inactivation of liver phosphorylase, the inactivation of muscle phosphorylase a (and its conversion to phosphorylase b), and the inactivation of muscle phosphorylase b kinase. Phosphatase activities against phosphoprotamine and muscle phosphorylase a copurify over the last three stages of purification. Phosphoprotamine inhibits phosphorylase phosphatase activity, and muscle phosphorylase a inhibits the dephosphorylation of phosphoprotamine. These results suggest that one enzyme possesses both phosphoprotamine phosphatase and phosphorylase phosphatase activities. The stimulation of phosphorylase phosphatase activity, but not of phosphoprotamine phosphatase activity, by caffeine and by glucose, suggests that the different activities of this phosphoprotein phosphatase may be regulated separately.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
DEAE–Sephadex®