Merck
2631605
Supelco
Inlet Liner, for Packed GC (unpacked)
pkg of 5 ea
About This Item
UNSPSC Code:
12000000
description
unpacked
packaging
pkg of 5 ea
L × O.D. × I.D.
112 mm × 6.0 mm × 3.0 mm
General description
  • length 112 mm × O.D. 6 mm × I.D. 3 mm
  • original Mfr part no. N610-1048
  • available unpacked or wool packed (for dirty samples)
·
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)