Documentos clave
42203-2
Millipore
Swing Bolts Kit, 6-tube T-Line
Seleccione un Tamaño
5 G
22.950 JPY
5 X 5 G
91.700 JPY
100 G
155.250 JPY
22.950 JPY
Precio de catálogo30.600 JPYAhorre 25 %
En stockDetalles
Seleccione un Tamaño
Cambiar Vistas
5 G
22.950 JPY
5 X 5 G
91.700 JPY
100 G
155.250 JPY
About This Item
UNSPSC Code:
41104903
eCl@ss:
32031602
22.950 JPY
Precio de catálogo30.600 JPYAhorre 25 %
En stockDetalles
Comparar elementos similares
Ver comparación completa
Mostrar Diferencias
1 of 4
Este artículo
AV44019HPA049000HPA065634
Quality Level
100
Quality Level
100
Quality Level
100
Quality Level
100
antibody form
affinity isolated antibody
antibody form
affinity isolated antibody
antibody form
affinity isolated antibody
antibody form
affinity isolated antibody
conjugate
unconjugated
conjugate
unconjugated
conjugate
unconjugated
conjugate
unconjugated
product line
Prestige Antibodies® Powered by Atlas Antibodies
product line
-
product line
Prestige Antibodies® Powered by Atlas Antibodies
product line
Prestige Antibodies® Powered by Atlas Antibodies
form
buffered aqueous glycerol solution
form
buffered aqueous solution
form
buffered aqueous glycerol solution
form
buffered aqueous glycerol solution
Certificados de análisis (COA)
¿Necesita un COA de muestra?
Esto es un Certificado de análisis (COA) de muestra y es posible que no represente un lote de fabricación reciente para este producto específico.