Documentos clave
CAD6332
1-way threaded end adapter (NPT)
FLL to 1/8-27 (NPT) thread (plated brass)
Seleccione un Tamaño
10 EA
MXP 7,943.00
MXP 7,943.00
Fecha estimada de envío23 de abril de 2025
Seleccione un Tamaño
Cambiar Vistas
10 EA
MXP 7,943.00
About This Item
Código UNSPSC:
41121815
NACRES:
NB.03
MXP 7,943.00
Fecha estimada de envío23 de abril de 2025
envase
pack of 10 ea
fabricante / nombre comercial
Cadence Science 6332
Descripción general
  • Plated brass or 316 stainless steel
  • Meets ISO 594 specifications
  • Final assembly water tested at 45 PSI
Female Luer Lock (FLL) - contains a tabbed hub for secure fitting to a threaded male Luer lock.
Micro-Mate® Female Luer Lock (MMFLL) - designed and manufactured to reduce the stripping of threads which occurs when metal adapters are mated with plastic male Luer lock adapters. Uniquely designed to facilitate easy cleaning and maintenance.
Male Luer Lock (MLL) - male Luer slip with a threaded sheath for a secure fitting to a female Luer lock.
Male Luer (ML) - male Luer slip that fits with a female Luer.
Información legal
Micro-Mate is a registered trademark of Cadence Science
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)