Keratin
Keratin is the primary fibrous structural protein component of hair, skin and nails. Keratin intermediate filaments are also a cytoskeletal component of desmosome cellular junctions. Keratins are highly cross-linked proteins typically containing α-helix and β-sheet motifs and are high in glycine and alanine content. Typically keratin exists as a heterodimeric paring of Type I, low MW acidic peptides and Type II high MW, neutral or basic peptides. At least 30 different genes code for keratin proteins.
Keratin Products | back to Structural Proteins |