Merck
HomePrimary Cell CulturePrimary Human Hepatocytes for In Vitro Testing
Primary Human Hepatocytes for In Vitro Testing
Human Hepatocytes are valuable and instrumental for in vitro testing. They have become the “gold standard” for metabolism, clearance, hepatotoxicity, and drug-drug interaction.
In order to best meet the specific needs of a wide variety of study types, we offer different hepatocyte products derived from human liver tissue. Our wide selection of lots that can be used for a variety of applications, including, but not
limited to:
  • Drug metabolism studies including stability, clearance, and species comparison
  • Induction and toxicity studies
  • Drug transporter assays (uptake and efflux)
Day 5 image of a selected lot of cryoplateable primary human hepatocytes at 10X magnification.
Figure 1. Day 5 image of a selected lot of cryoplateable primary human hepatocytes at 10X magnification.
Each batch of hepatocytes are characterized with batch-specific functionality and clinical data to best permit lot selections and reservations:
  • Cell viability and yield per vial
  • Morphological integrity and attachment efficiency
  • Monolayer confluency
  • Phase I and phase II drug metabolizing enzyme activities
  • CYP induction when applicable
  • Additional functional testing can be performed upon request for specific cell culture applications
  • A comprehensive Certificate of Analysis (CoA) with relevant donor demographics, BMI, pre-mortem liver function lab values, and medical history
Click here to get a complete list of currently available hepatocyte lots from LifeNet Health®.
LifeNet Health Primary Human Hepatocytes