Merck
HomeGene Expression & SilencingBiodegradable Nanoparticle
Biodegradable Nanoparticle
Preparation of Biodegradable Nanoparticles and Their Use in Transfection
Jaspreet K. Vasir and Vinod Labhasetwar
This protocol was adapted from "Biodegradable Nanoparticles," Chapter 56, in Gene Transfer: Delivery and Expression of DNA and RNA (eds. Friedmann and Rossi). Cold Spring Harbor Laboratory Press, Cold Spring Harbor, NY, USA, 2007.
Products Available for this Protocol
Sorry, an unexpected error has occurred
Network error: Failed to fetch
Product Association Disclaimer: The products listed for this specific protocol were selected either to match or to supplement the products listed within the actual protocol. Our products/reagents have been qualified for usage but may not have been validated for this specific application. Please refer to the detailed product description on the usage of specific products of interest.
ABSTRACT
Biodegradable nanoparticles (NPs) are colloidal particles with a gene of interest encapsulated inside a polymeric matrix. They are typically 100 nm in diameter, and are formulated using FDA-approved, biodegradable, biocompatible polymers such as poly(D,L-lactide-co-glycolide) (PLGA) or polylactide (PLA). The NPs are taken up by cells via an endocytic process, and the encapsulated plasmid DNA entrapped in NPs is protected from degradation by both extra- and intracellular nucleases. It is released slowly, sustaining gene delivery and gene expression. In contrast, higher but transient gene expression is observed with lipid- or polymer-based complexes where most of the delivered DNA is available quickly for transfection. Thus, unlike other nonviral gene delivery systems, NPs thus constitute a sustained gene expression vector. Sustained gene expression is advantageous, especially when the half-life of the expressed protein is very low or when chronic gene delivery is required for therapeutic efficacy. This protocol describes a method for nanoencapsulation of DNA and the subsequent use of NPs for transfection.