HPLC Analysis of Fatty Acid Methyl Esters (FAMES) on SUPELCOSIL™ LC-18
Materials
analytical column
Product No.
Description
Pricing
CONDITIONS
column
SUPELCOSIL LC-18, 25 cm × 4.6 mm I.D., 5 μm particles (58298)
mobile phase
[A] acetonitrile; [B] acetone; (59.0:41.0, v/v, A:B)
flow rate
1 mL/min
detector
RI
injection
10 μL of 9% C8 to C18 saturated and unsaturated FAMEs in mobile phase
Description
Legal Information
SUPELCOSIL is a trademark of Sigma-Aldrich Co. LLC