Merck
GC Analysis of Fluorocarbons on a Packed Column (5% Fluorocol on 60/80 Carbopack B)
GC Analysis of Fluorocarbons on a Packed Column (5% Fluorocol™ on 60/80 Carbopack™ B) suitable for GC
CONDITIONS
column
10’ x 1/8” SP® alloy, general configuration, packed with 5% Fluorcol on 60/80 Carbopack B
oven
50 °C
detector
FID
carrier gas
nitrogen, 30 mL/min
injection
10 μL synthetic mixture (approximately 20% each compound)
Description
Analysis Note
A packed GC column using Fluorcol stationary phase is also useful for fluorocarbons. The polarity of this column provides an alternate elution pattern compared to the Alumina chloride PLOT column chromatogram on the previous slide.
Legal Information
Carbopack is a trademark of Merck KGaA, Darmstadt, Germany
Fluorcol is a trademark of Sigma-Aldrich Co. LLC
SP is a registered trademark of Sigma-Aldrich Co. LLC