US EPA Method 8270: GC Analysis of Semivolatiles on SLB®-5ms (20 m x 0.18 mm I.D., 0.18 μm), Fast GC Analysis
Materials
analytical column
Product No.
Description
Pricing
CONDITIONS
column
SLB-5ms, 20 m x 0.18 mm I.D., 0.18 μm (28564-U)
oven
40 °C (0.7 min), 55 °C/min to 240 °C, 28 °C/min to 330 °C (2 min)
inj. temp.
250 °C
MSD interface
330 °C
scan range
m/z = 40-450
carrier gas
helium, 40 cm/sec, constant
injection
0.5 μL, 10:1 split
liner
2 mm I.D., fast FocusLiner™ liner with taper (2879501-U)
sample
80 component semivolatile standard at 50 ppm plus 6 internal standards (at 40 ppm) in methylene chloride
Description
Legal Information
FocusLiner is a trademark of SGE Analytical Science Pty Ltd.
SLB is a registered trademark of Merck KGaA, Darmstadt, Germany