- Sauvagine cross-links to the second extracellular loop of the corticotropin-releasing factor type 1 receptor.
Sauvagine cross-links to the second extracellular loop of the corticotropin-releasing factor type 1 receptor.
Contact sites between the corticotropin-releasing factor receptor type 1 (CRFR1), the sauvagine (SVG) radioligands [Tyr(0),Gln(1)]SVG ((125)I-YQS) and [Tyr(0),Gln(1), Leu(17)]SVG ((125)I-YQLS) were examined. (125)I-YQLS or (125)I-YQS was cross-linked to CRFR1 using the chemical cross-linker, disuccinimidyl suberate (DSS), which cross-links the epsilon amino groups of lysine residues that have a molecular distance of 11.4 A. DSS specifically and efficiently cross-linked (125)I-YQLS and (125)I-YQS to CRFR1. CRFR1 contains 5 putative extracellular lysine residues (Lys(110), Lys(111), Lys(113), Lys(257), and Lys(262)) that can cross-link to the 4 lysine residues (Lys(16), Lys(22), Lys(25), and Lys(27)) of the radioligands. Identification of the CNBr-cleaved fragments of CRFR1 cross-linked to (125)I-YQLS or (125)I-YQS established that the second extracellular loop of CRFR1 cross-links to Lys(16) of YQS. Additionally, site-directed mutagenesis (changing Lys to Arg in CRFR1 individually and in combination) revealed that Lys(257) in the second extracellular loop of CRFR1 is an important cross-linking site. In conclusion, it was shown that in SVG-bound CRFR1, Lys(257) of CRFR1 lies in close proximity (11.4 A) to Lys(16) of SVG.