Merck
  • Cannabinoids Regulate the Diameter of Pericyte-Containing Retinal Capillaries in Rats.
Cannabinoids Regulate the Diameter of Pericyte-Containing Retinal Capillaries in Rats.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology (2017-10-24)
Yuan Zong, Xujiao Zhou, Jingyi Cheng, Jian Yu, Jihong Wu, Chunhui Jiang
ABSTRACT
Cannabinoids are vasoactive substances that act as key regulators of arterial tone in the blood vessels supplying peripheral tissues and the central nervous system. We therefore investigated the effect of cannabinoids on retinal capillaries and pericytes. The effects of cannabinoids on capillary diameters were determined using an ex vivo whole-mount rat retinal model. Western blotting, quantitative PCR, and immunohistochemistry were performed to explore the underlying mechanism. Endogenous cannabinoid 2-arachidonoylglycerol and anandamide and exogenous cannabinoid (R-(+)-WIN55212-2) dilated the noradrenaline-precontracted capillaries in a concentration-dependent manner (1 µM to 0.1 mM). The extent of vasorelaxation was positively correlated with changes in pericyte width. The effects of R-(+)-WIN55212-2 on vasorelaxation and pericyte width were inhibited by a cannabinoid receptor type-1 (CB1) antagonist, AM251 or rimonabant (SR141716A), the nitric oxide synthase inhibitor l-NAME, and the guanylate cyclase inhibitor ODQ. They were also abolished by the removal of the endothelium, but not by the cannabinoid receptor-2 antagonist SR144528, the endothelial cannabinoid receptor antagonist O-1918, or the cyclooxygenase inhibitor indomethacin. The exogenous cannabinoid R-(+)-WIN55212-2 promotes the vasorelaxation of pericyte-containing rat retinal capillaries. This effect of R-(+)-WIN55212-2 is dependent on CB1 and the nitric oxide-cyclic guanosine monophosphate pathway, and requires an intact endothelium.
Sigma-Aldrich
AM251, >98% (HPLC), solid
Sigma-Aldrich
Arachidonylethanolamide, ≥97.0% (TLC), oil
Sigma-Aldrich
CHAPS 100 mM solution
Sigma-Aldrich
1H-[1,2,4]Oxadiazolo[4,3-a]quinoxalin-1-one, powder