- Green-fabrication of gold nanomaterials using Staphylococcus warneri from Sundarbans estuary: an effective recyclable nanocatalyst for degrading nitro aromatic pollutants.
Green-fabrication of gold nanomaterials using Staphylococcus warneri from Sundarbans estuary: an effective recyclable nanocatalyst for degrading nitro aromatic pollutants.
Environmental science and pollution research international (2017-11-11)
Sudip Nag, Arnab Pramanik, Dhrubajyoti Chattopadhyay, Maitree Bhattacharyya
PMID29124636
ABSTRACT
Microbial synthesis of gold nanoparticles (GNPs) has attracted considerable attention in recent times due to their exceptional capability for the bioremediation of industrial wastes and also for the treatment of wastewater. A bacterial strain Staphylococcus warneri, isolated from the estuarine mangroves of Sundarbans region produced highly stable GNPs by reducing hydrogen auric chloride (HAucl