Merck
  • Chemokine Signaling Facilitates Early-Stage Breast Cancer Survival and Invasion through Fibroblast-Dependent Mechanisms.
Chemokine Signaling Facilitates Early-Stage Breast Cancer Survival and Invasion through Fibroblast-Dependent Mechanisms.
Molecular cancer research : MCR (2017-11-15)
Gage Brummer, Diana S Acevedo, Qingting Hu, Mike Portsche, Wei Bin Fang, Min Yao, Brandon Zinda, Megan Myers, Nehemiah Alvarez, Patrick Fields, Yan Hong, Fariba Behbod, Nikki Cheng
ABSTRACT
Ductal carcinoma in situ (DCIS) is the most common form of breast cancer, with 50,000 cases diagnosed every year in the United States. Overtreatment and undertreatment remain significant clinical challenges in patient care. Identifying key mechanisms associated with DCIS progression could uncover new biomarkers to better predict patient prognosis and improve guided treatment. Chemokines are small soluble molecules that regulate cellular homing through molecular gradients. CCL2-mediated recruitment of CCR2+ macrophages are a well-established mechanism for metastatic progression. Although the CCL2/CCR2 pathway is a therapeutic target of interest, little is known about the role of CCR2 expression in breast cancer. Here, using a mammary intraductal injection (MIND) model to mimic DCIS formation, the role of CCR2 was explored in minimally invasive SUM225 and highly invasive DCIS.com breast cancer cells. CCR2 overexpression increased SUM225 breast cancer survival and invasion associated with accumulation of CCL2 expressing fibroblasts. CCR2-deficient DCIS.com breast cancer cells formed fewer invasive lesions with fewer CCL2+ fibroblasts. Cografting CCL2-deficient fibroblasts with DCIS.com breast cancer cells in the subrenal capsule model inhibited tumor invasion and survival associated with decreased expression of aldehyde dehydrogenase (ALDH1), a proinvasive factor, and decreased expression of HTRA2, a proapoptotic serine protease. Through data mining analysis, high expression of CCR2 and ALDH1 and low HTRA2 expression were correlated with poor prognosis of breast cancer patients.Implications: This study demonstrates that CCR2 overexpression in breast cancer drives early-stage breast cancer progression through stromal-dependent expression of CCL2 with important insight into prognosis and treatment of DCIS. Mol Cancer Res; 16(2); 296-308. ©2017 AACR.