Merck
  • 5-Azacytidine specifically inhibits the NIH-3T3 PCD process induced by TNF-alpha and cycloheximide via affecting BCL-XL.
5-Azacytidine specifically inhibits the NIH-3T3 PCD process induced by TNF-alpha and cycloheximide via affecting BCL-XL.
Journal of cellular biochemistry (2017-08-05)
Qing Wang, Pu Wang, Hong Zhou, Yan Hu, Chengshen Xie, Fei Gao, Ningjie Ma, Haoli Hou, Hao Zhang, Lijia Li
ABSTRACT
DNA methylation plays a crucial role in lots of biological processes and cancer. 5-azacytidine (5-AC), a DNA methylation inhibitor, has been used as a potential chemotherapeutic agent for cancer. In this study, we used 5-AC treatment to investigate whether DNA methylation was involved in regulation of programmed cell death (PCD) in mouse embryo fibroblast NIH-3T3 cells which could undergo PCD after treatment with TNF-α and cycloheximide (CHX). The results showed that the genomic DNA of NIH-3T3 cells was hypermethylated during PCD induced by TNF-α and CHX, and 5-AC might prevent this PCD process. However, treatment with the other three DNA methylation inhibitors, 5-aza-deoxycytidine, 6-thioguanine and RG108, did not interfere with the NIH-3T3 cell PCD process. Additionally, knockdown of DNMT1 did not affect the apoptosis process. The present results and observations indicated that 5-AC specifically inhibited the NIH-3T3 apoptosis process via a genomic DNA methylation-independent pathway. During the TNF-α and CHX-inducing apoptosis process, the PCD related BCL-2 family proteins were significantly down-regulated. Furthermore, after the small interference RNA-mediated knockdown of BCL-XL, one of the BCL-2 family proteins, 5-AC did not inhibit the apoptosis process, suggesting that 5-AC inhibited the PCD process induced by TNF-α and CHX by affecting the anti-apoptotic protein BCL-XL.
Sigma-Aldrich
RG108, ≥98% (HPLC), powder