Merck
  • A highly sensitive impedimetric aptasensor for the selective detection of acetamiprid and atrazine based on microwires formed by platinum nanoparticles.
A highly sensitive impedimetric aptasensor for the selective detection of acetamiprid and atrazine based on microwires formed by platinum nanoparticles.
Biosensors & bioelectronics (2017-11-03)
L Madianos, G Tsekenis, E Skotadis, L Patsiouras, D Tsoukalas
ABSTRACT
A novel impedimetric biosensor was developed for the detection of the two extensively used pesticides, acetamiprid and atrazine. By employing the sputtering and e-beam lithography techniques, platinum nanoparticles (Pt NPs) were deposited in a bridge-like arrangement, in between interdigitated electrodes (IDEs). The resulting Pt NP microwires were chemically functionalized to allow the covalent immobilization of aptamers against the two target analytes onto the sensor surfaces. The biosensing platform facilitated charge transfer through the microwire-bridged IDEs, while upon analyte binding to the immobilized aptamers electron transfer was hindered, resulting in an increase of the electrochemical cell's impedance. The combination of Pt NPs microwires and aptamers allowed the sensitive and highly selective detection of acetamiprid with a linear range of response in the range of 10pM to 100nM with a limit of detection (LoD) at 1pM, and of atrazine with a linear range of responses from 100pM to 1μM and a LoD at 10pM respectively. Its performance was tested against a number of other commonly used pesticides as well as in real water samples.