Merck
  • Emerging functions of c-kit and its ligand stem cell factor in dendritic cells: regulators of T cell differentiation.
Emerging functions of c-kit and its ligand stem cell factor in dendritic cells: regulators of T cell differentiation.
Cell cycle (Georgetown, Tex.) (2008-09-13)
Prabir Ray, Nandini Krishnamoorthy, Anuradha Ray
ABSTRACT
The receptor tyrosine kinase, c-kit, and its ligand, stem cell factor (SCF), function in a diverse range of biological functions. The role of c-kit in the maintenance and survival of hematopoietic stem cells and of mast cells is well recognized. c-kit also plays an important role in melanogenesis, erythropoiesis and spermatogenesis. Recent work from our laboratory highlights an important role of c-kit in the regulation of expression of two molecules in dendritic cells (DCs), interleukin-6 (IL-6) and Jagged-2 (a ligand of Notch), which are known to regulate T helper cell differentiation. Our study shows that induction of c-kit expression and its signaling in DCs promotes Th2 and Th17 responses but not Th1 response. c-kit inhibition by imatinib mesylate (Gleevec) in DCs was previously shown to promote natural killer cell activation which may be due to dampening of IL-6 production by the DCs. Since dysregulation of c-kit function has been associated with various disease states including cancer, in this perspective we have focused on known and novel functions of c-kit to include molecules such as IL-6 and Notch that were not previously recognized to be within the purview of c-kit biology. We have also reviewed the differential expression pattern of SCF and c-kit on various cell types and its variation during development or pathology. The recognition of previously unappreciated roles for c-kit will provide better insights into its function within and beyond the immune system and pave the way for developing better therapeutic strategies.
Sigma-Aldrich
Stem Cell Factor human, SCF, recombinant, expressed in HEK 293 cells, suitable for cell culture
Sigma-Aldrich
Stem Cell Factor from mouse, SCF, recombinant, expressed in E. coli, powder, suitable for cell culture