- Adsorption-desorption and hysteresis phenomenon of tebuconazole in Colombian agricultural soils: Experimental assays and mathematical approaches.
Adsorption-desorption and hysteresis phenomenon of tebuconazole in Colombian agricultural soils: Experimental assays and mathematical approaches.
Chemosphere (2017-10-13)
Carmen S Mosquera-Vivas, María J Martinez, Glenda García-Santos, Jairo A Guerrero-Dallos
PMID29024884
ABSTRACT
The adsorption-desorption, hysteresis phenomenon, and leachability of tebuconazole were studied for Inceptisol and Histosol soils at the surface (0-10 cm) and in the subsurface (40-50 cm) of an agricultural region from Colombia by the batch-equilibrium method and mathematical approaches. The experimental K
MATERIALS