Merck
  • Development of correction formula for field potential duration of human induced pluripotent stem cell-derived cardiomyocytes sheets.
Development of correction formula for field potential duration of human induced pluripotent stem cell-derived cardiomyocytes sheets.
Journal of pharmacological sciences (2017-09-21)
Hiroko Izumi-Nakaseko, Yasunari Kanda, Yuji Nakamura, Mihoko Hagiwara-Nagasawa, Takeshi Wada, Kentaro Ando, Atsuhiko T Naito, Yuko Sekino, Atsushi Sugiyama
ABSTRACT
Human induced pluripotent stem cell-derived cardiomyocytes (hiPSC-CMs) have been used in many studies to assess proarrhythmic risks of chemical compounds. In those studies, field potential durations (FPD) of hiPSC-CMs have been corrected by clinically used Fridericia's and/or Bazett's formulae, however, the rationale for the use of these formulae has not been well established. In the present study, we developed a correction formula for experiments using hiPSC-CMs. First, we analyzed the effect of beating rate on FPD in the hiPSC-CMs sheets with electrical stimuli and a HCN channel inhibitor zatebradine. Next, we examined the relationship between the electrophysiological properties and the expression levels of ion channel genes in the cell sheets. Zatebradine slowed the beating rate and allowed to analyze FPD changes at various pacing cycle lengths. Rate-dependent change in the repolarization period was smaller in the cell sheets than that reported on the human hearts, which can be partly explained by lower gene expression level of hKCNJ2 and hKCNE1. Thus, non-linear equation for correcting FPD in the cell sheet; FPDc = FPD/RR
Sigma-Aldrich
Zatebradine hydrochloride, ≥98% (HPLC), powder