- Evaluation of the efficacy of
Evaluation of the efficacy of
Experimental and therapeutic medicine (2017-11-08)
Yoko Sawada, Atsushi Sugimoto, Takehito Hananouchi, Norimasa Sato, Isao Nagaoka
PMID29109757
ABSTRACT
The aim of the present study was to assess the efficacy and safety of the oral administration of