Merck
  • The NEIL1 G83D germline DNA glycosylase variant induces genomic instability and cellular transformation.
The NEIL1 G83D germline DNA glycosylase variant induces genomic instability and cellular transformation.
Oncotarget (2017-11-22)
Heather A Galick, Carolyn G Marsden, Scott Kathe, Julie A Dragon, Lindsay Volk, Antonia A Nemec, Susan S Wallace, Aishwarya Prakash, Sylvie Doublié, Joann B Sweasy
ABSTRACT
Base excision repair (BER) is a key genome maintenance pathway. The NEIL1 DNA glycosylase recognizes oxidized bases, and likely removes damage in advance of the replication fork. The rs5745906 SNP of the NEIL1 gene is a rare human germline variant that encodes the NEIL1 G83D protein, which is devoid of DNA glycosylase activity. Here we show that expression of G83D NEIL1 in MCF10A immortalized but non-transformed mammary epithelial cells leads to replication fork stress. Upon treatment with hydrogen peroxide, we observe increased levels of stalled replication forks in cells expressing G83D NEIL1 versus cells expressing the wild-type (WT) protein. Double-strand breaks (DSBs) arise in G83D-expressing cells during the S and G2/M phases of the cell cycle. Interestingly, these breaks result in genomic instability in the form of high levels of chromosomal aberrations and micronuclei. Cells expressing G83D also grow in an anchorage independent manner, suggesting that the genomic instability results in a carcinogenic phenotype. Our results are consistent with the idea that an inability to remove oxidative damage in an efficient manner at the replication fork leads to genomic instability and mutagenesis. We suggest that individuals who harbor the G83D NEIL1 variant face an increased risk for human cancer.
Sigma-Aldrich
Triton X-100, laboratory grade
Ouabain, European Pharmacopoeia (EP) Reference Standard