Merck
  • Involvement of miR-451 in resistance to paclitaxel by regulating YWHAZ in breast cancer.
Involvement of miR-451 in resistance to paclitaxel by regulating YWHAZ in breast cancer.
Cell death & disease (2017-10-06)
Wenrui Wang, Lingyu Zhang, Yangyang Wang, Yongxing Ding, Tiantian Chen, Yueyue Wang, Haifeng Wang, Yu Li, Kecai Duan, Sulian Chen, Qingling Yang, Changjie Chen
ABSTRACT
MicroRNAs (miRNAs) have been identified as major post-transcriptional regulators of the initiation and progression of human cancers, including breast cancer. However, the detail role of miR-451 has not been fully elucidated in breast cancer. In this study, we aimed to investigate the biological role and molecular mechanisms of miR-451 in drug resistance in breast cancer cell lines and in xenograft model. We show that miR-451 is decreased in human breast cancer specimens and in paclitaxel-resistant (PR) cells. Ectopic expression of miR-451 could inhibit the cell migration and invasion, promoted apoptosis, induced cell-cycle arrest Furthermore, tyrosine3-monooxygenase/tryptophan5-monooxygenase activation protein zeta (YWHAZ) was identified as a direct target of miR-451. Remarkably, the expression of YWHAZ is inversely correlated with the level of miR-451 in human breast cancer samples. Co-treatment with miR-451 mimics and YWHAZ-siRNA significantly enhanced YWHAZ knockdown in both SKBR3/PR and MCF-7/PR cells Moreover, miR-451 markedly inhibited expression of β-catenin via YWHAZ and subsequently inhibited downstream gene cyclin D1, c-Myc expression. The results of xenograft model in vivo showed that intratumor injection of miR-451 agomir induced a tumor-suppressive effect in SKBR3/PR drug-resistant xenograft model. Taken together, our findings suggested that miR-451 might be considered as important and potential target in paclitaxel-resistant breast cancer treatment.
Sigma-Aldrich
MISSION® esiRNA, targeting human YWHAZ