Merck
  • Control of reactivity through chemical order in very small RuRe nanoparticles.
Control of reactivity through chemical order in very small RuRe nanoparticles.
Dalton transactions (Cambridge, England : 2003) (2017-10-27)
Tuğçe Ayvalı, Pier-Francesco Fazzini, Pierre Lecante, Alvaro Mayoral, Karine Philippot, Bruno Chaudret
ABSTRACT
The choice of suitable organometallic precursors, [Re