Merck
  • Simultaneous Enantioselective Determination of the Chiral Fungicide Prothioconazole and Its Major Chiral Metabolite Prothioconazole-Desthio in Food and Environmental Samples by Ultraperformance Liquid Chromatography-Tandem Mass Spectrometry.
Simultaneous Enantioselective Determination of the Chiral Fungicide Prothioconazole and Its Major Chiral Metabolite Prothioconazole-Desthio in Food and Environmental Samples by Ultraperformance Liquid Chromatography-Tandem Mass Spectrometry.
Journal of agricultural and food chemistry (2017-08-29)
Zhaoxian Zhang, Qing Zhang, Beibei Gao, Gaozhang Gou, Lianshan Li, Haiyan Shi, Minghua Wang
ABSTRACT
An efficient and sensitive chiral analytical method was established for the determination of the chiral fungicide prothioconazole and its major chiral metabolite prothioconazole-desthio in agricultural and environmental samples using ultraperformance liquid chromatography-tandem mass spectrometry. The optical rotation and absolute configuration of enantiomers were identified by optical rotation detector and electronic circular dichroism spectra. The elution order of prothioconazole and its chiral metabolite enantiomers was R-(+)-prothioconazole-desthio, S-(-)-prothioconazole-desthio, R-(-)-prothioconazole, and S-(+)-prothioconazole. The mean recoveries from the samples was 71.8-102.0% with intraday relative standard deviations (RSDs) of 0.3-11.9% and interday RSDs of 0.9-10.6%. The formation of prothioconazole-desthio was studied in soil under field conditions and enantioselective degradation was observed for chiral prothioconazole. Remarkable enantioselective degradation was observed: R-prothioconazole degraded preferentially with EF values from 0.48 to 0.37. Although prothioconazole-desthio is the most remarkably bioactive metabolite, no obvious enantioselective behavior was observed in soil. These results may help to systematically evaluate prothioconazole and its metabolites in food and environmental safety.