Merck
  • Solvent-controlled leaving-group selectivity in aromatic nucleophilic substitution.
Solvent-controlled leaving-group selectivity in aromatic nucleophilic substitution.
Organic letters (2008-10-10)
Lukas Hintermann, Ritsuki Masuo, Keisuke Suzuki
ABSTRACT
A solvent-controlled inversion of leaving group ability allows selective access to either of two internal substitution products in S(N)Ar reactions of substrates with competing leaving groups. Application of this principle in a selective synthesis of the highly functionalized xanthone core of the antibiotic FD-594 is presented.