Merck
  • A Pt(0) complex with cyclic (alkyl)(amino)silylene and 1,3-divinyl-1,1,3,3-tetramethyldisiloxane ligands: synthesis, molecular structure, and catalytic hydrosilylation activity.
A Pt(0) complex with cyclic (alkyl)(amino)silylene and 1,3-divinyl-1,1,3,3-tetramethyldisiloxane ligands: synthesis, molecular structure, and catalytic hydrosilylation activity.
Dalton transactions (Cambridge, England : 2003) (2017-05-16)
Tomohiro Iimura, Naohiko Akasaka, Tomoyuki Kosai, Takeaki Iwamoto
ABSTRACT
A platinum(0) complex bearing a cyclic (alkyl)(amino)silylene and a 1,3-divinyl-1,1,3,3-tetramethyldisiloxane (DVTMS) was synthesized and isolated in the form of colorless crystals. The single-crystal X-ray diffraction analysis of this complex in combination with theoretical calculations indicated that the Pt→Si π-back-donation in this complex is weaker than that in the corresponding cyclic-dialkylsilylene-ligated Pt complex. The performance of this complex in the catalytic hydrosilylation of (Me
Sigma-Aldrich
N-Allyl-N,N-bis(trimethylsilyl)amine, 97%