Merck
  • Effect of aging and repeated intratracheal challenge on levels of cryptic and overt natural antibodies in poultry.
Effect of aging and repeated intratracheal challenge on levels of cryptic and overt natural antibodies in poultry.
Poultry science (2010-01-16)
T V L Berghof, G De Vries Reilingh, M G B Nieuwland, H K Parmentier
ABSTRACT
Natural antibodies (NAb) have been divided in 2 classes: overt and cryptic. Overt NAb can be detected in unfractionated normal sera of nonimmunized mammals and chickens. Cryptic NAb as described in mammals need an in vitro physical or biochemical treatment to be detected, which may reflect their biochemical modification in situ during inflammation or infection. We studied the effect of concurrent primary, secondary, and tertiary intratracheal (i.t.) challenges with lipopolysaccharide (LPS) and human serum albumin on levels of 2 cryptic NAb [i.e., NAb binding actin (ACT), or thyroglobulin (THYRO)] and levels of an overt NAb binding keyhole limpet hemocyanin (KLH). In addition, effect of aging of the birds on levels of NAb was taken into account. Presence and changes on the level of the cryptic antibodies directed to ACT and THYRO were expected after the i.t. challenges with LPS (and human serum albumin). However, levels of NAb binding ACT were only significantly enhanced by LPS after primary challenge, whereas levels of NAb binding THYRO were not affected by primary nor secondary challenges but were significantly decreased by LPS after the third immunization. On the other hand, no changes in the levels of overt NAb binding KLH were expected, but levels of NAb binding KLH were significantly enhanced after the primary and secondary challenges with LPS. Levels of all three NAb increased with aging, but the different challenges performed at 3 moments during aging did not significantly or consistently affect levels of the overt nor cryptic NAb. Our results suggest that chickens might react by overt NAb as well as cryptic NAb to an infection or inflammation rather than by cryptic NAb only. The relation between various types of NAb and i.t. immunization with antigen and LPS and aging is discussed.
Sigma-Aldrich
Actin from bovine muscle, lyophilized powder