Merck
  • Enhancement of Oxidative Desulfurization Performance over UiO-66(Zr) by Titanium Ion Exchange.
Enhancement of Oxidative Desulfurization Performance over UiO-66(Zr) by Titanium Ion Exchange.
Chemphyschem : a European journal of chemical physics and physical chemistry (2017-04-25)
Gan Ye, Hui Qi, Xiaolin Li, Kunyue Leng, Yinyong Sun, Wei Xu
ABSTRACT
Oxidative desulfurization is considered to be one of the most promising methods for producing ultra-low-sulfur fuels because it can effectively remove refractory sulfur-containing aromatic compounds under mild conditions. In this work, the oxidative desulfurization performance over UiO-66(Zr) is greatly enhanced by Ti ion exchange. This strategy is not only efficient for UiO-66(Zr) with crystal defects but also for UiO-66(Zr) with high crystallinity. In particular, the performance of UiO-66(Zr) with high crystallinity in the oxidative desulfurization of dibenzothiophene can be improved more than 11-fold, which can be mainly attributed to the introduction of active Ti sites.
Sigma-Aldrich
4,6-Dimethyldibenzothiophene, 97%