Merck
  • (Pro)renin receptor (ATP6AP2) depletion arrests As4.1 cells in the G0/G1 phase thereby increasing formation of primary cilia.
(Pro)renin receptor (ATP6AP2) depletion arrests As4.1 cells in the G0/G1 phase thereby increasing formation of primary cilia.
Journal of cellular and molecular medicine (2017-02-20)
Heike Wanka, Philipp Lutze, Doreen Staar, Barbara Peters, Anica Morch, Lukas Vogel, Ravi Kumar Chilukoti, Georg Homuth, Jaroslaw Sczodrok, Inga Bäumgen, Jörg Peters
ABSTRACT
The (pro)renin receptor [(P)RR, ATP6AP2] is a multifunctional transmembrane protein that activates local renin-angiotensin systems, but also interacts with Wnt pathways and vacuolar H
Sigma-Aldrich
MISSION® esiRNA, targeting human ATP6AP2