Merck
  • Activation of adenosine A2A receptor signaling regulates the expression of cytokines associated with immunologic dysfunction in BTBR T
Activation of adenosine A2A receptor signaling regulates the expression of cytokines associated with immunologic dysfunction in BTBR T
Molecular and cellular neurosciences (2017-05-04)
Mushtaq A Ansari, Sabry M Attia, Ahmed Nadeem, Saleh A Bakheet, Mohammad Raish, Tajdar H Khan, Othman A Al-Shabanah, Sheikh F Ahmad
ABSTRACT
Autism spectrum disorder (ASD) is neurodevelopmental disorders characterized by stereotypical repetitive behavior, impaired social interaction, and deficits in communication. The BTBR T
Sigma-Aldrich
SCH 58261, ≥98% (HPLC), solid