Merck
  • Critical reappraisal confirms that Mitofusin 2 is an endoplasmic reticulum-mitochondria tether.
Critical reappraisal confirms that Mitofusin 2 is an endoplasmic reticulum-mitochondria tether.
Proceedings of the National Academy of Sciences of the United States of America (2016-09-21)
Deborah Naon, Marta Zaninello, Marta Giacomello, Tatiana Varanita, Francesca Grespi, Sowmya Lakshminaranayan, Annalisa Serafini, Martina Semenzato, Stephanie Herkenne, Maria Isabel Hernández-Alvarez, Antonio Zorzano, Diego De Stefani, Gerald W Dorn, Luca Scorrano
ABSTRACT
The discovery of the multiple roles of mitochondria-endoplasmic reticulum (ER) juxtaposition in cell biology often relied upon the exploitation of Mitofusin (Mfn) 2 as an ER-mitochondria tether. However, this established Mfn2 function was recently questioned, calling for a critical re-evaluation of Mfn2's role in ER-mitochondria cross-talk. Electron microscopy and fluorescence-based probes of organelle proximity confirmed that ER-mitochondria juxtaposition was reduced by constitutive or acute Mfn2 deletion. Functionally, mitochondrial uptake of Ca
Sigma-Aldrich
Anti-MICU1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution