Merck
  • Triggering Receptor Expressed on Myeloid Cells 2, a Novel Regulator of Immunocyte Phenotypes, Confers Neuroprotection by Relieving Neuroinflammation.
Triggering Receptor Expressed on Myeloid Cells 2, a Novel Regulator of Immunocyte Phenotypes, Confers Neuroprotection by Relieving Neuroinflammation.
Anesthesiology (2017-04-12)
Qian Zhai, Feng Li, Xiyao Chen, Ji Jia, Sisi Sun, Dandan Zhou, Lei Ma, Tao Jiang, Fuhai Bai, Lize Xiong, Qiang Wang
ABSTRACT
Microglia can not only detrimentally augment secondary injury but also potentially promote recovery. However, the mechanism underlying the regulation of microglial phenotypes after stroke remains unclear. Mice were subjected to middle cerebral artery occlusion for 60 min. At 3 days after reperfusion, the effects of activation and suppression of triggering receptor expressed on myeloid cells 2 on immunocyte phenotypes (n = 5), neurobehavioral scores (n = 7), infarct volumes (n = 8), and neuronal apoptosis (n = 7) were analyzed. In vitro, cultured microglia were exposed to oxygen-glucose deprivation for 4 h. Inflammatory cytokines, cellular viability (n = 8), neuronal apoptosis (n = 7), and triggering receptor expressed on myeloid cells 2 expression (n = 5) were evaluated in the presence or absence of triggering receptor expressed on myeloid cell-specific small interfering RNA or triggering receptor expressed on myeloid cells 2 overexpression lentivirus. Triggering receptor expressed on myeloid cells 2 expression in the ischemic penumbra peaked at 3 days after ischemia-reperfusion injury (4.4 ± 0.1-fold, P = 0.0004) and was enhanced in interleukin-4/interleukin-13-treated microglia in vitro (1.7 ± 0.2-fold, P = 0.0119). After oxygen-glucose deprivation, triggering receptor expressed on myeloid cells 2 conferred neuroprotection by regulating the phenotypic conversion of microglia and inflammatory cytokine release. Intraperitoneal administration of triggering receptor expressed on myeloid cells 2 agonist heat shock protein 60 or unilateral delivery of a recombinant triggering receptor expressed on myeloid cells 2 lentivirus into the cerebral ventricle induced a significant neuroprotective effect in mice (apoptotic neurons decreased to 31.3 ± 7.6%; infarct volume decreased to 44.9 ± 5.3%). All values are presented as the mean ± SD. Activation or up-regulation of triggering receptor expressed on myeloid cells 2 promoted the phenotypic conversion of microglia and decreased the number of apoptotic neurons. Our study suggests that triggering receptor expressed on myeloid cells 2 is a novel regulator of microglial phenotypes and may be a potential therapeutic target for stroke.
Sigma-Aldrich
TREM2 human, recombinant, expressed in E. coli, ≥90% (SDS-PAGE)