Merck
  • Crystalline magnesium chloride-electron donor complexes: new support materials for Ziegler-Natta catalysts.
Crystalline magnesium chloride-electron donor complexes: new support materials for Ziegler-Natta catalysts.
Dalton transactions (Cambridge, England : 2003) (2017-03-18)
V H Nissinen, I O Koshevoy, T T Pakkanen
ABSTRACT
In this study, crystalline magnesium chloride-electron donor complexes were prepared by recrystallization of δ-MgCl
Sigma-Aldrich
N,N′-Diethylethylenediamine, 95%
Sigma-Aldrich
1-Chlorobutane, ReagentPlus®, 99%