- Nicotine-based surface active ionic liquids: Synthesis, self-assembly and cytotoxicity studies.
Nicotine-based surface active ionic liquids: Synthesis, self-assembly and cytotoxicity studies.
Journal of colloid and interface science (2017-02-27)
Gurbir Singh, Raman Kamboj, Venus Singh Mithu, Vinay Chauhan, Taranjeet Kaur, Gurcharan Kaur, Sukhprit Singh, Tejwant Singh Kang
PMID28236691
ABSTRACT
New ester-functionalized surface active ionic liquids (SAILs) based on nicotine, [C