Merck
  • In vitro profiling of toxicity and endocrine disrupting effects of bisphenol analogues by employing MCF-7 cells and two-hybrid yeast bioassay.
In vitro profiling of toxicity and endocrine disrupting effects of bisphenol analogues by employing MCF-7 cells and two-hybrid yeast bioassay.
Environmental toxicology (2016-02-27)
Bingli Lei, Jie Xu, Wei Peng, Yu Wen, Xiangying Zeng, Zhiqiang Yu, Yipei Wang, Tian Chen
ABSTRACT
The potentially adverse health implications of bisphenol A (BPA) have led to increasing use of alternative bisphenols (BPs). However, little is known about the toxicity of alternative BPs. In this study, the cytotoxicity, genotoxicity, intracellular ROS formation, and Ca
Sigma-Aldrich
L-Thyronine
Sigma-Aldrich
4,4′-(Hexafluoroisopropylidene)diphenol, 97%
Supelco
Tetrachlorobisphenol A, analytical standard