Merck
  • Urinary Polycyclic Aromatic Hydrocarbon Metabolites and Human Semen Quality in China.
Urinary Polycyclic Aromatic Hydrocarbon Metabolites and Human Semen Quality in China.
Environmental science & technology (2016-12-15)
Pan Yang, Yi-Xin Wang, Ying-Jun Chen, Li Sun, Jin Li, Chong Liu, Zhen Huang, Wen-Qing Lu, Qiang Zeng
ABSTRACT
Toxicological studies have demonstrated that polycyclic aromatic hydrocarbons (PAHs) exposure impairs male reproductive health. However, the epidemiological evidence is limited and discordant. Our goal was to investigate the relationship between PAH exposures and human semen quality. We analyzed 12 urinary metabolites of PAHs from 933 men who sought semen quality analysis in an infertility clinic in Wuhan, China. Associations with semen quality were assessed using a multivariable linear regression. Restricted cubic splines were used to explore the dose-response relationships between urinary metabolites of PAHs and semen quality. We observed inverse associations between urinary 1-hydroxynaphthalene (1-OHNa) and sperm count, sperm concentration, and percentage of normal morphology (all p for trends <0.05) as well as between urinary ∑OHNa (sum of 1-OHNa and 2-OHNa) and sperm concentration (p for trend =0.04). Additionally, we found inverse associations between urinary 9-hydroxyphenanthrene (9-OHPh) and semen volume and sperm straight-line velocity (both p for trends <0.05) as well as between urinary ∑OHPh (sum of 1-, 2-, 3-, 4-, and 9-OHPh) and sperm count (p for trend =0.04). These dose-response relationships were further confirmed in the curves of the restricted cubic splines. Our data suggest that exposure to naphthalene and phenanthrene is related to decreased semen quality. Our results contribute to the growing body of evidence regarding the widespread exposure to PAHs and the detriment to male reproductive function.
Sigma-Aldrich
2-Hydroxyfluorene, 98%