Merck
  • The scaffolding function of the RLTPR protein explains its essential role for CD28 co-stimulation in mouse and human T cells.
The scaffolding function of the RLTPR protein explains its essential role for CD28 co-stimulation in mouse and human T cells.
The Journal of experimental medicine (2016-11-05)
Romain Roncagalli, Margot Cucchetti, Nicolas Jarmuzynski, Claude Grégoire, Elise Bergot, Stéphane Audebert, Emilie Baudelet, Marisa Goncalves Menoita, Anais Joachim, Stéphane Durand, Miloslav Suchanek, Frédéric Fiore, Lichen Zhang, Yinming Liang, Luc Camoin, Marie Malissen, Bernard Malissen
ABSTRACT
The RLTPR cytosolic protein, also known as CARMIL2, is essential for CD28 co-stimulation in mice, but its importance in human T cells and mode of action remain elusive. Here, using affinity purification followed by mass spectrometry analysis, we showed that RLTPR acts as a scaffold, bridging CD28 to the CARD11/CARMA1 cytosolic adaptor and to the NF-κB signaling pathway, and identified proteins not found before within the CD28 signaling pathway. We further demonstrated that RLTPR is essential for CD28 co-stimulation in human T cells and that its noncanonical pleckstrin-homology domain, leucine-rich repeat domain, and proline-rich region were mandatory for that task. Although RLTPR is thought to function as an actin-uncapping protein, this property was dispensable for CD28 co-stimulation in both mouse and human. Our findings suggest that the scaffolding role of RLTPR predominates during CD28 co-stimulation and underpins the similar function of RLTPR in human and mouse T cells. Along that line, the lack of functional RLTPR molecules impeded the differentiation toward Th1 and Th17 fates of both human and mouse CD4+ T cells. RLTPR was also expressed in both human and mouse B cells. In the mouse, RLTPR did not play, however, any detectable role in BCR-mediated signaling and T cell-independent B cell responses.
Sigma-Aldrich
N-[Tris(hydroxymethyl)methyl]acrylamide, contains ≤7% KCl, 93%