Merck
  • NNZ-2566: a Gly-Pro-Glu analogue with neuroprotective efficacy in a rat model of acute focal stroke.
NNZ-2566: a Gly-Pro-Glu analogue with neuroprotective efficacy in a rat model of acute focal stroke.
Journal of the neurological sciences (2009-01-23)
Michael J Bickerdike, Gregory B Thomas, David C Batchelor, Ernest S Sirimanne, Wing Leong, Hai Lin, Frank Sieg, Jingyuan Wen, Margaret A Brimble, Paul W Harris, Peter D Gluckman
ABSTRACT
The N-terminal cleavage product of human insulin-like growth factor-1 (IGF-1) in the brain is the tripeptide molecule Glypromate (Gly-Pro-Glu). Glypromate has demonstrated neuroprotective effects in numerous in vitro and in vivo models of brain injury and is in clinical trials for the prevention of cognitive impairment following cardiac surgery. NNZ-2566 is a structural analogue of Glypromate, resulting from alpha-methylation of the proline moiety, which has improved the elimination half-life and oral bioavailability over the parent peptide. In vivo, NNZ-2566 reduces injury size in rats subjected to focal stroke. An intravenous infusion of NNZ-2566 of 4 h duration (3-10 mg/kg/h), initiated 3 h after endothelin-induced middle-cerebral artery constriction, significantly reduced infarct area as assessed on day 5. Neuroprotective efficacy in the MCAO model was also observed following oral administration of the drug (30-60 mg/kg), when formulated as a microemulsion. In vitro, NNZ-2566 significantly attenuates apoptotic cell death in primary striatal cultures, suggesting attenuation of apoptosis is one mechanism of action underlying its neuroprotective effects. NNZ-2566 is currently in clinical trials for the treatment of cognitive deficits following traumatic brain injury, and these data further support the development of the drug as a neuroprotective agent for acute brain injury.
Sigma-Aldrich
Trofinetide Trifluoroacetate, ≥95% (HPLC)