Merck
  • A novel application of carbon dots for colorimetric determination of fenitrothion insecticide based on the microextraction method.
A novel application of carbon dots for colorimetric determination of fenitrothion insecticide based on the microextraction method.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2016-09-03)
A Larki
ABSTRACT
In this work, the intrinsic colorimetric property of carbon dots (CDs) was utilized for the determination of fenitrothion by applying dispersive liquid-liquid microextraction (DLLME) method. Label free CDs are extracted into carbon tetrachloride via assistance of trioctylmethylammonium chloride (Aliquat 336), which also acts as a disperser agent in this technique. The enriched CDs show an absorption signal at 365nm, which increases in the presence of fenitrothion. The absorbance increase of CDs in organic phase was used as an analytical signal for the determination of fenitrothion. The synthesized CDs were characterized by UV-visible, fluorescence, Fourier transform infrared (FT-IR) and transmission electron microscopic (TEM). Under the optimized conditions, beer's law was obeyed in the range of 1.0-250.0ngmL-1 of fenitrothion with the limit of detection for fenitrothion was 0.2ngmL-1. The relative standard deviation for eight replicate measurements of fenitrothion at concentrations of 25 and 100ngmL-1 were calculated to be 1.5 and 3.7%, respectively. The proposed method was successfully applied in the determination of fenitrothion in water samples with satisfactory results.
Sigma-Aldrich
Methyltrioctylammonium chloride, ≥97.0% (AT)