Merck
Bioinspired nanoscale materials for biomedical and energy applications.
Journal of the Royal Society, Interface (2014-04-18)
Priyanka Bhattacharya, Dan Du, Yuehe Lin
ABSTRACT
The demand for green, affordable and environmentally sustainable materials has encouraged scientists in different fields to draw inspiration from nature in developing materials with unique properties such as miniaturization, hierarchical organization and adaptability. Together with the exceptional properties of nanomaterials, over the past century, the field of bioinspired nanomaterials has taken huge leaps. While on the one hand, the sophistication of hierarchical structures endows biological systems with multi-functionality, the synthetic control on the creation of nanomaterials enables the design of materials with specific functionalities. The aim of this review is to provide a comprehensive, up-to-date overview of the field of bioinspired nanomaterials, which we have broadly categorized into biotemplates and biomimics. We discuss the application of bioinspired nanomaterials as biotemplates in catalysis, nanomedicine, immunoassays and in energy, drawing attention to novel materials such as protein cages. Furthermore, the applications of bioinspired materials in tissue engineering and biomineralization are also discussed.