MilliporeSigma
  • DDTs and other organochlorine pesticides in tissues of four bird species from the Rift Valley region, Ethiopia.
DDTs and other organochlorine pesticides in tissues of four bird species from the Rift Valley region, Ethiopia.
The Science of the total environment (2016-08-20)
Yared Beyene Yohannes, Yoshinori Ikenaka, Shouta M M Nakayama, Hazuki Mizukawa, Mayumi Ishizuka
ABSTRACT
Despite the presence of a wide variety and number of birds, there is exceedingly little data on organochlorine pesticide (OCP) residues in birds inhabiting in Africa. In the present study, concentrations of dichlorodiphenyltrichloroethanes (DDTs), hexachlorocyclohexanes, drins, cyclodienes, and hexachlorobenzene were measured in liver, kidney, heart and brain of 4 bird species from the Rift Valley region, Ethiopia. Indoor residual spraying of DDT for malaria vector control, and indiscriminate and illegal use of pesticides underline the relevance of this study. Levels of ΣOCPs ranged from 1.87 to 4586ng/g wet weight, and the scavenger bird species Leptoptilos crumeniferus had the highest level in liver. In all tissues, contamination profiles of OCPs within the species were similar, with DDTs≫other OCPs. Among the DDTs, p,p'-DDE was the most abundant compound and had significantly a higher burden in all tissues. The risk characterization demonstrated potential risks to the studied birds associated with DDE exposure. Maximum hepatic levels of p,p'-DDE exceeded the levels reported to trigger adverse effects. The detection of p,p'-DDT in all bird tissues suggests the release of fresh DDT to the environment. This is the first study to assay OCPs in different tissues of birds from the Ethiopian Rift Valley region, and henceforth the data will serve as a reference data for future studies.