Merck
  • 2,2,2-trifluoroethyl formate: a versatile and selective reagent for the formylation of alcohols, amines, and N-hydroxylamines.
2,2,2-trifluoroethyl formate: a versatile and selective reagent for the formylation of alcohols, amines, and N-hydroxylamines.
Organic letters (2002-01-05)
David R Hill, Chi-Nung Hsiao, Ravi Kurukulasuriya, Steven J Wittenberger
ABSTRACT
[reaction: see text] Treatment of a variety of alcohols, amines, and N-hydroxylamines with 2,2,2-trifluoroethyl formate gave the corresponding formylated adducts in high yields.
Sigma-Aldrich
2,2,2-Trifluoroethyl formate, 95%