Merck
  • Mechanisms of PDGF siRNA-mediated inhibition of bone cancer pain in the spinal cord.
Mechanisms of PDGF siRNA-mediated inhibition of bone cancer pain in the spinal cord.
Scientific reports (2016-06-11)
Yang Xu, Jia Liu, Mu He, Ran Liu, Visar Belegu, Ping Dai, Wei Liu, Wei Wang, Qing-Jie Xia, Fei-Fei Shang, Chao-Zhi Luo, Xue Zhou, Su Liu, JohnW McDonald, Jin Liu, Yun-Xia Zuo, Fei Liu, Ting-Hua Wang
ABSTRACT
Patients with tumors that metastasize to bone frequently suffer from debilitating pain, and effective therapies for treating bone cancer are lacking. This study employed a novel strategy in which herpes simplex virus (HSV) carrying a small interfering RNA (siRNA) targeting platelet-derived growth factor (PDGF) was used to alleviate bone cancer pain. HSV carrying PDGF siRNA was established and intrathecally injected into the cavum subarachnoidale of animals suffering from bone cancer pain and animals in the negative group. Sensory function was assessed by measuring thermal and mechanical hyperalgesia. The mechanism by which PDGF regulates pain was also investigated by comparing the differential expression of pPDGFRα/β and phosphorylated ERK and AKT. Thermal and mechanical hyperalgesia developed in the rats with bone cancer pain, and these effects were accompanied by bone destruction in the tibia. Intrathecal injection of PDGF siRNA and morphine reversed thermal and mechanical hyperalgesia in rats with bone cancer pain. In addition, we observed attenuated astrocyte hypertrophy, down-regulated pPDGFRα/β levels, reduced levels of the neurochemical SP, a reduction in CGRP fibers and changes in pERK/ERK and pAKT/AKT ratios. These results demonstrate that PDGF siRNA can effectively treat pain induced by bone cancer by blocking the AKT-ERK signaling pathway.